Anti-SAT2 Mouse Monoclonal Antibody [clone: 2B3]

Proveedor: OriGene
CF504632
ORIGCF504632EA 786 EUR
ORIGCF504632
Anti-SAT2 Mouse Monoclonal Antibody [clone: 2B3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: SAT2
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR