Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1D4]

Proveedor: OriGene
CF504626
ORIGCF504626EA 764 EUR
ORIGCF504626
Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: C17orf37
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR