Anti-CRYZL1 Mouse Monoclonal Antibody [clone: 1B3]

Proveedor: OriGene
CF504617
ORIGCF504617EA 764 EUR
ORIGCF504617
Anti-CRYZL1 Mouse Monoclonal Antibody [clone: 1B3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: CRYZL1
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR