Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1E4]

Proveedor: OriGene
CF504609
ORIGCF504609EA 764 EUR
ORIGCF504609
Anti-C17orf37 Mouse Monoclonal Antibody [clone: 1E4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: C17orf37
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR