Anti-CENPH Mouse Monoclonal Antibody [clone: 2G4]

Proveedor: OriGene
CF503872
ORIGCF503872EA 764 EUR
ORIGCF503872
Anti-CENPH Mouse Monoclonal Antibody [clone: 2G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CENPH
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR