Anti-HARS2 Mouse Monoclonal Antibody [clone: 6F9]

Proveedor: OriGene
CF503866
ORIGCF503866EA 764 EUR
ORIGCF503866
Anti-HARS2 Mouse Monoclonal Antibody [clone: 6F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HARS2
Clonality: Monoclonal
Clone: 6F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR