Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: 3H10]

Proveedor: OriGene
CF503845
ORIGCF503845EA 764 EUR
ORIGCF503845
Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: 3H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LDLRAP1
Clonality: Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR