Anti-MDM4 Mouse Monoclonal Antibody [clone: 4B10]

Proveedor: OriGene
CF505733
ORIGCF505733EA 764 EUR
ORIGCF505733
Anti-MDM4 Mouse Monoclonal Antibody [clone: 4B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MDM4
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR