Anti-PDIA3 Mouse Monoclonal Antibody [clone: 3D2]

Proveedor: OriGene
CF504990
ORIGCF504990EA 764 EUR
ORIGCF504990
Anti-PDIA3 Mouse Monoclonal Antibody [clone: 3D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR