Anti-POFUT2 Mouse Monoclonal Antibody [clone: 3C12]

Proveedor: OriGene
CF504983
ORIGCF504983EA 764 EUR
ORIGCF504983
Anti-POFUT2 Mouse Monoclonal Antibody [clone: 3C12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: POFUT2
Clonality: Monoclonal
Clone: 3C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR