Anti-LRRC50 Mouse Monoclonal Antibody [clone: 3E2]

Proveedor: OriGene
CF504577
ORIGCF504577EA 764 EUR
ORIGCF504577
Anti-LRRC50 Mouse Monoclonal Antibody [clone: 3E2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LRRC50
Clonality: Monoclonal
Clone: 3E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR