Anti-NUDT18 Mouse Monoclonal Antibody [clone: 1A8]

Proveedor: OriGene
CF503831
ORIGCF503831EA 786 EUR
ORIGCF503831
Anti-NUDT18 Mouse Monoclonal Antibody [clone: 1A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NUDT18
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR