Anti-MDM4 Mouse Monoclonal Antibody [clone: 4G5]

Proveedor: OriGene
CF505723
ORIGCF505723EA 764 EUR
ORIGCF505723
Anti-MDM4 Mouse Monoclonal Antibody [clone: 4G5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MDM4
Clonality: Monoclonal
Clone: 4G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR