Anti-ATXN7L1 Mouse Monoclonal Antibody [clone: 4B12]

Proveedor: OriGene
CF505710
ORIGCF505710EA 786 EUR
ORIGCF505710
Anti-ATXN7L1 Mouse Monoclonal Antibody [clone: 4B12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ATXN7L1
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR