Anti-TMEFF2 Mouse Monoclonal Antibody [clone: 1E4]

Proveedor: OriGene
ORIGCF504556EA 764 EUR
ORIGCF504556
Anti-TMEFF2 Mouse Monoclonal Antibody [clone: 1E4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEFF2
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR