Anti-LRRC50 Mouse Monoclonal Antibody [clone: 4E4]

Proveedor: OriGene
CF504535
ORIGCF504535EA 764 EUR
ORIGCF504535
Anti-LRRC50 Mouse Monoclonal Antibody [clone: 4E4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: LRRC50
Clonality: Monoclonal
Clone: 4E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR