Anti-UNG Mouse Monoclonal Antibody [clone: 1G2]

Proveedor: OriGene
CF503755
ORIGCF503755EA 764 EUR
ORIGCF503755
Anti-UNG Mouse Monoclonal Antibody [clone: 1G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: UNG
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR