Anti-NUBPL Mouse Monoclonal Antibody [clone: 5D5]

Proveedor: OriGene
CF503744
ORIGCF503744EA 764 EUR
ORIGCF503744
Anti-NUBPL Mouse Monoclonal Antibody [clone: 5D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NUBPL
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR