Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 1A8]

Proveedor: OriGene
CF503741
ORIGCF503741EA 786 EUR
ORIGCF503741
Anti-TBC1D21 Mouse Monoclonal Antibody [clone: 1A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TBC1D21
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR