Anti-PRMT2 Mouse Monoclonal Antibody [clone: 1A2]

Proveedor: OriGene
CF503740
ORIGCF503740EA 786 EUR
ORIGCF503740
Anti-PRMT2 Mouse Monoclonal Antibody [clone: 1A2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PRMT2
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR