Anti-PDE6G Mouse Monoclonal Antibody [clone: 1E11]

Proveedor: OriGene
CF503739
ORIGCF503739EA 764 EUR
ORIGCF503739
Anti-PDE6G Mouse Monoclonal Antibody [clone: 1E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE6G
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR