Anti-PDE6G Mouse Monoclonal Antibody [clone: 2F5]

Proveedor: OriGene
CF503736
ORIGCF503736EA 764 EUR
ORIGCF503736
Anti-PDE6G Mouse Monoclonal Antibody [clone: 2F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: PDE6G
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR