Anti-NUBPL Mouse Monoclonal Antibody [clone: 1C5]

Proveedor: OriGene
CF503726
ORIGCF503726EA 764 EUR
ORIGCF503726
Anti-NUBPL Mouse Monoclonal Antibody [clone: 1C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NUBPL
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR