Anti-SEC14L2 Mouse Monoclonal Antibody [clone: 4H2]

Proveedor: OriGene
CF503723
ORIGCF503723EA 764 EUR
ORIGCF503723
Anti-SEC14L2 Mouse Monoclonal Antibody [clone: 4H2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SEC14L2
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR