Anti-ASCC1 Mouse Monoclonal Antibody [clone: 1E2]

Proveedor: OriGene
CF503720
ORIGCF503720EA 764 EUR
ORIGCF503720
Anti-ASCC1 Mouse Monoclonal Antibody [clone: 1E2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ASCC1
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR