Anti-ADAMTS8 Mouse Monoclonal Antibody [clone: 2H5]

Proveedor: OriGene
ORIGCF505684EA 764 EUR
ORIGCF505684
Anti-ADAMTS8 Mouse Monoclonal Antibody [clone: 2H5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: ADAMTS8
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR