Anti-HRAS Mouse Monoclonal Antibody [clone: 1F2]

Proveedor: OriGene
CF505665
ORIGCF505665EA 764 EUR
ORIGCF505665
Anti-HRAS Mouse Monoclonal Antibody [clone: 1F2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: HRAS
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR