Anti-COMMD1 Mouse Monoclonal Antibody [clone: 2E2]

Proveedor: OriGene
CF504899
ORIGCF504899EA 764 EUR
ORIGCF504899
Anti-COMMD1 Mouse Monoclonal Antibody [clone: 2E2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: COMMD1
Clonality: Monoclonal
Clone: 2E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR