Anti-COMMD1 Mouse Monoclonal Antibody [clone: 3B3]

Proveedor: OriGene
CF504889
ORIGCF504889EA 764 EUR
ORIGCF504889
Anti-COMMD1 Mouse Monoclonal Antibody [clone: 3B3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: COMMD1
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR