Anti-MLF2 Mouse Monoclonal Antibody [clone: 1G1]

Proveedor: OriGene
CF504876
ORIGCF504876EA 786 EUR
ORIGCF504876
Anti-MLF2 Mouse Monoclonal Antibody [clone: 1G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MLF2
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR