Anti-CST3 Mouse Monoclonal Antibody [clone: 1A6]

Proveedor: OriGene
CF504494
ORIGCF504494EA 764 EUR
ORIGCF504494
Anti-CST3 Mouse Monoclonal Antibody [clone: 1A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CST3
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR