Anti-CNOT4 Mouse Monoclonal Antibody [clone: 2A5]

Proveedor: OriGene
CF504473
ORIGCF504473EA 764 EUR
ORIGCF504473
Anti-CNOT4 Mouse Monoclonal Antibody [clone: 2A5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CNOT4
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR