Anti-NIF3L1 Mouse Monoclonal Antibody [clone: 5A7]

Proveedor: OriGene
CF503680
ORIGCF503680EA 764 EUR
ORIGCF503680
Anti-NIF3L1 Mouse Monoclonal Antibody [clone: 5A7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NIF3L1
Clonality: Monoclonal
Clone: 5A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR