Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 3D10]

Proveedor: OriGene
CF505642
ORIGCF505642EA 764 EUR
ORIGCF505642
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 3D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR