Anti-ETS2 Mouse Monoclonal Antibody [clone: 3C4]

Proveedor: OriGene
CF505621
ORIGCF505621EA 786 EUR
ORIGCF505621
Anti-ETS2 Mouse Monoclonal Antibody [clone: 3C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ETS2
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR