Anti-POGK Mouse Monoclonal Antibody [clone: 4C8]

Proveedor: OriGene
CF505608
ORIGCF505608EA 764 EUR
ORIGCF505608
Anti-POGK Mouse Monoclonal Antibody [clone: 4C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: POGK
Clonality: Monoclonal
Clone: 4C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR