Anti-DPP3 Mouse Monoclonal Antibody [clone: 2G8]

Proveedor: OriGene
CF503291
ORIGCF503291EA 786 EUR
ORIGCF503291
Anti-DPP3 Mouse Monoclonal Antibody [clone: 2G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP3
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR