Anti-SIRT5 Mouse Monoclonal Antibody [clone: 2A11]

Proveedor: OriGene
CF503282
ORIGCF503282EA 786 EUR
ORIGCF503282
Anti-SIRT5 Mouse Monoclonal Antibody [clone: 2A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: SIRT5
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR