Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1H3]

Proveedor: OriGene
CF503271
ORIGCF503271EA 764 EUR
ORIGCF503271
Anti-PSMD2 Mouse Monoclonal Antibody [clone: 1H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PSMD2
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR