Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 2D3]

Proveedor: OriGene
CF503269
ORIGCF503269EA 764 EUR
ORIGCF503269
Anti-ALDH3A2 Mouse Monoclonal Antibody [clone: 2D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: ALDH3A2
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR