Anti-SSR1 Mouse Monoclonal Antibody [clone: 5D5]

Proveedor: OriGene
CF503265
ORIGCF503265EA 786 EUR
ORIGCF503265
Anti-SSR1 Mouse Monoclonal Antibody [clone: 5D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: SSR1
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR