Anti-ERCC4 Mouse Monoclonal Antibody [clone: 4E11]

Proveedor: OriGene
CF503261
ORIGCF503261EA 764 EUR
ORIGCF503261
Anti-ERCC4 Mouse Monoclonal Antibody [clone: 4E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC4
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR