Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 5A9]

Proveedor: OriGene
CF500503
ORIGCF500503EA 764 EUR
ORIGCF500503
Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 5A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SLC7A8
Clonality: Monoclonal
Clone: 5A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR