Anti-CTAG1B Mouse Monoclonal Antibody [clone: 3F10]

Proveedor: OriGene
CF500498
ORIGCF500498EA 764 EUR
ORIGCF500498
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 3F10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:50-100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 3F10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR