Anti-MAPK1 Mouse Monoclonal Antibody [clone: 4C2]

Proveedor: OriGene
CF500485
ORIGCF500485EA 764 EUR
ORIGCF500485
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 4C2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR