Anti-RPA2 Mouse Monoclonal Antibody [clone: 7F4]

Proveedor: OriGene
CF500812
ORIGCF500812EA 764 EUR
ORIGCF500812
Anti-RPA2 Mouse Monoclonal Antibody [clone: 7F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: RPA2
Clonality: Monoclonal
Clone: 7F4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR