Anti-LEMD3 Mouse Monoclonal Antibody [clone: 6C10]

Proveedor: OriGene
CF500794
ORIGCF500794EA 786 EUR
ORIGCF500794
Anti-LEMD3 Mouse Monoclonal Antibody [clone: 6C10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: LEMD3
Clonality: Monoclonal
Clone: 6C10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR