Anti-BTRC Mouse Monoclonal Antibody [clone: 3D5]

Proveedor: OriGene
CF502464
ORIGCF502464EA 764 EUR
ORIGCF502464
Anti-BTRC Mouse Monoclonal Antibody [clone: 3D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: BTRC
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR