Anti-NRBP1 Mouse Monoclonal Antibody [clone: 2C8]

Proveedor: OriGene
CF500457
ORIGCF500457EA 764 EUR
ORIGCF500457
Anti-NRBP1 Mouse Monoclonal Antibody [clone: 2C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR