Anti-SHPRH Mouse Monoclonal Antibody [clone: 5A8]

Proveedor: OriGene
CF501445
ORIGCF501445EA 786 EUR
ORIGCF501445
Anti-SHPRH Mouse Monoclonal Antibody [clone: 5A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SHPRH
Clonality: Monoclonal
Clone: 5A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR